Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290274.1 | NP_001277203.1 | 7 | 1889 | ||
NM_153319.3 | NP_695231.3 | 7 | 1889 | |||
GenBank mRNA | AF461135.1 | - | 7 | 1293 | ||
AK129277.1 | - | 2 | 129 | |||
BC057638.1 | - | 7 | 1344 | 2631191 | ||
CK392840.1 | - | 4 | 335 |