Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030245.3 | NP_084521.1 | 4 | 547 | ||
GenBank mRNA | AK013599.1 | - | 4 | 314 | ||
AK151132.1 | - | 4 | 409 | |||
BC002307.1 | - | 4 | 349 | 3592046 | ||
BC027337.1 | - | 4 | 414 | 3995355 |