Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030245.3 | NP_084521.1 | 5 | 637 | ||
GenBank mRNA | AK013599.1 | - | 5 | 404 | ||
AK151132.1 | - | 5 | 499 | |||
BC002307.1 | - | 5 | 439 | 3592046 | ||
BC027337.1 | - | 5 | 504 | 3995355 |