Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138583.2 | NP_613049.2 | 5 | 432 | ||
GenBank mRNA | AK089239.1 | - | 5 | 414 | ||
AK133588.1 | - | 5 | 415 | |||
AK168595.1 | - | 4 | 411 | |||
BC005445.1 | - | 5 | 493 | 3496749 | ||
X74504.1 | - | 4 | 405 |