Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138583.2 | NP_613049.2 | 7 | 593 | ||
GenBank mRNA | AK089239.1 | - | 7 | 575 | ||
AK133588.1 | - | 7 | 576 | |||
AK168595.1 | - | 6 | 572 | |||
AK199175.1 | - | 2 | 135 | |||
BC005445.1 | - | 7 | 673 | 3496749 | ||
X74504.1 | - | 6 | 566 |