Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cherp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138585.3 | NP_613051.3 | 10 | 1644 | ||
GenBank mRNA | AF536541.1 | - | 10 | 1546 | ||
AK049765.1 | - | 10 | 1554 | |||
AK144328.1 | - | 10 | 1564 | |||
AK171907.1 | - | 3 | 620 | |||
BC032191.1 | - | 10 | 1562 | 5322208 |