Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004137.3 | NP_004128.1 | 4 | 1289 | ||
GenBank mRNA | U25138.1 | - | 4 | 1005 | ||
U42600.1 | - | 4 | 899 | |||
U61536.1 | - | 4 | 950 |