Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253843.1 | NP_001240772.1 | 2 | 340 | ||
NM_134129.4 | NP_598890.1 | 2 | 340 | |||
GenBank mRNA | AB222602.1 | - | 2 | 125 | ||
AF251503.1 | - | 2 | 305 | |||
AK047693.1 | - | 2 | 158 | |||
AK088769.1 | - | 2 | 325 | |||
AK164681.1 | - | 2 | 328 | |||
BC004070.1 | - | 2 | 279 | 3591050 |