Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178603.4 | NP_848718.1 | 2 | 248 | ||
GenBank mRNA | AK002646.1 | - | 2 | 236 | ||
AK002686.1 | - | 2 | 250 | |||
AK007405.1 | - | 2 | 245 | |||
AK031229.1 | - | 2 | 249 | |||
AK049421.1 | - | 2 | 238 | |||
AK075787.1 | - | 2 | 230 | |||
BC021318.1 | - | 2 | 259 | 5035369 |