Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145353.2 | NP_663328.2 | 3 | 523 | ||
GenBank mRNA | AF197931.1 | - | 2 | 283 | ||
AK011822.1 | - | 3 | 510 | |||
AK042644.1 | - | 2 | 195 | |||
AK149957.1 | - | 3 | 356 | |||
AK177708.1 | - | 2 | 94 | |||
BC011161.1 | - | 3 | 484 | |||
BC019384.1 | - | 3 | 504 | 5123736 | ||
BC100575.1 | - | 3 | 455 | 6432108 |