Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145353.2 | NP_663328.2 | 8 | 1043 | ||
GenBank mRNA | AK011822.1 | - | 8 | 1030 | ||
AK042644.1 | - | 7 | 715 | |||
AK149957.1 | - | 8 | 876 | |||
BC011161.1 | - | 8 | 1004 | |||
BC019384.1 | - | 8 | 1024 | 5123736 | ||
BC100575.1 | - | 7 | 1111 | 6432108 |