Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138596.2 | NP_613062.1 | 3 | 325 | ||
GenBank mRNA | AK003862.1 | - | 3 | 319 | ||
AK013992.1 | - | 3 | 329 | |||
AK156498.1 | - | 3 | 334 | |||
AK215905.1 | - | 3 | 173 | |||
BC048638.1 | - | 3 | 318 | 6771841 | ||
BC056438.1 | - | 3 | 314 | 6818638 | ||
BC059231.1 | - | 2 | 2807 | |||
BC089312.1 | - | 2 | 1870 |