Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170855.1 | NP_001164326.1 | 8 | 1707 | ||
NM_178872.4 | NP_849203.2 | 8 | 1708 | |||
GenBank mRNA | AB103063.1 | - | 8 | 1502 | ||
AY251389.1 | - | 8 | 1465 | |||
BC058106.1 | - | 8 | 1626 | 6836197 |