Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053014.3 | NP_443747.2 | 6 | 721 | ||
GenBank mRNA | AB377215.1 | - | 4 | 537 | ||
AK008965.1 | - | 6 | 877 | |||
AK015906.1 | - | 5 | 649 | |||
AK030607.1 | - | 4 | 513 | |||
AK075715.1 | - | 6 | 721 | |||
AK076414.1 | - | 5 | 690 | |||
AK159110.1 | - | 5 | 961 | |||
AY167588.1 | - | 4 | 559 | |||
BC033444.1 | - | 4 | 399 | |||
BC052382.1 | - | 5 | 612 | 5703571 | ||
BC058519.1 | - | 5 | 611 | 5703571 |