Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm70a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138599.5 | NP_613065.2 | 11 | 1747 | ||
GenBank mRNA | AK012084.1 | - | 11 | 1748 | ||
AK083586.1 | - | 11 | 1748 | |||
AK122356.1 | - | 1680 | ||||
AK145458.1 | - | 11 | 1760 | |||
AK150429.1 | - | 11 | 1748 | |||
AK161720.1 | - | 11 | 1737 | |||
AK169809.1 | - | 11 | 1709 | |||
AK207500.1 | - | 2 | 154 | |||
BC057096.1 | - | 11 | 1748 | 5709110 | ||
BC139420.1 | - | 11 | 1720 | 8862442 | ||
BC139421.1 | - | 11 | 1720 | 8862443 |