Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037938.2 | NP_001033027.2 | 2 | 438 | ||
NM_030686.2 | NP_109611.1 | 2 | 438 | |||
GenBank mRNA | AB045132.1 | - | 2 | 350 | ||
AK018788.1 | - | 2 | 321 | |||
AK146988.1 | - | 2 | 318 | |||
BC003484.1 | - | 2 | 273 | 2811468 | ||
BC054361.1 | - | 2 | 399 | 5008299 |