Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013797.5 | NP_038825.1 | 3 | 586 | ||
GenBank mRNA | AB031813.1 | - | 3 | 427 | ||
AF148218.1 | - | 3 | 410 | |||
AF223067.1 | - | 3 | 420 | |||
AK041125.1 | - | 3 | 434 | |||
AK165360.1 | - | 3 | 441 | |||
AY195868.1 | - | 3 | 419 | |||
AY195869.1 | - | 3 | 419 | |||
AY243579.1 | - | 3 | 419 | |||
BC041147.1 | - | 3 | 369 | 4975986 |