Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znhit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013859.2 | NP_038887.2 | 1 | 1011 | ||
GenBank mRNA | AF119498.1 | - | 1 | 1020 | ||
AK086863.1 | - | 1 | 1011 | |||
AK164029.1 | - | 1 | 1009 | |||
BC019440.1 | - | 1 | 997 | 5125922 | ||
BC092256.1 | - | 1 | 1026 | 5102003 |