Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013863.5 | NP_038891.4 | 4 | 1630 | ||
GenBank mRNA | AB041583.1 | - | 4 | 1597 | ||
AF130471.1 | - | 4 | 1363 | |||
AK007414.1 | - | 4 | 1441 | |||
AK016510.1 | - | 4 | 1437 | |||
AK036675.1 | - | 4 | 1439 | |||
AK171734.1 | - | 4 | 1572 | |||
BC145765.1 | - | 4 | 1543 | 40131068 |