Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145959.1 | NP_001139431.1 | 3 | 260 | ||
NM_013864.2 | NP_038892.1 | 4 | 302 | |||
GenBank mRNA | AB033921.1 | - | 4 | 165 | ||
AK154597.1 | - | 4 | 170 | |||
AK158900.1 | - | 3 | 169 | |||
AK168011.1 | - | 3 | 127 | |||
BB848819.1 | - | 3 | 259 | |||
BC012963.1 | - | 4 | 169 | 4211421 |