Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNQ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004518.5 | NP_004509.2 | 5, 6 | 1037 | ||
NM_172106.2 | NP_742104.1 | 5, 6 | 1037 | |||
NM_172107.3 | NP_742105.1 | 5, 6 | 1037 | |||
NM_172108.4 | NP_742106.1 | 5, 6 | 1037 | |||
NM_172109.2 | NP_742107.1 | 5, 6 | 1037 | |||
XM_011528810.1 | XP_011527112.1 | 5, 6 | 949 | |||
XM_011528811.1 | XP_011527113.1 | 5, 6 | 949 | |||
XM_017027841.1 | XP_016883330.1 | 5, 6 | 949 | |||
XM_017027842.1 | XP_016883331.1 | 5, 6 | 949 | |||
XM_017027843.1 | XP_016883332.1 | 5, 6 | 842 | |||
XM_017027844.1 | XP_016883333.1 | 5, 6 | 949 | |||
GenBank mRNA | AF033348.1 | - | 5, 6 | 950 | ||
AF074247.1 | - | 5, 6 | 998 | |||
AF110020.1 | - | 5, 6 | 869 | |||
AK293727.1 | - | 5, 6 | 949 | |||
BC000699.2 | - | 5, 6 | 866 | |||
BT007043.1 | - | 5, 6 | 823 | |||
D82346.1 | - | 5, 6 | 1000 | |||
Y15065.1 | - | 5, 6 | 865 |