Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159284.1 | NP_001152756.1 | 12 | 1909 | ||
NM_001284427.1 | NP_001271356.1 | 11 | 1858 | |||
NM_001284428.1 | NP_001271357.1 | 12 | 1966 | |||
NM_001284429.1 | NP_001271358.1 | 12 | 1911 | |||
NM_013870.3 | NP_038898.2 | 11 | 1858 | |||
GenBank mRNA | AF064236.1 | - | 11 | 1805 | ||
AJ010305.1 | - | 11 | 1805 | |||
AK030932.1 | - | 11 | 1859 | |||
AK083947.1 | - | 1 | 112 | |||
AK085412.1 | - | 12 | 1873 | |||
AK138898.1 | - | 12 | 1923 | |||
AK160492.1 | - | 2 | 313 | |||
AK166385.1 | - | 12 | 1926 | |||
AK170516.1 | - | 11 | 1818 | |||
BC010599.1 | - | 11 | 1799 | 3709078 | ||
BC066192.1 | - | 12 | 1784 | 30433616 | ||
BC069836.1 | - | 12 | 1784 | 30456568 | ||
BC069839.1 | - | 12 | 1784 | 30431675 |