Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013879.2 | NP_038907.1 | 5 | 540 | ||
GenBank mRNA | AB079604.1 | - | 5 | 479 | ||
AF169152.1 | - | 5 | 540 | |||
AF169153.1 | - | 4 | 360 | |||
AK141231.1 | - | 5 | 454 | |||
KJ364651.1 | - | 4 | 978 |