Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtse1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168672.1 | NP_001162143.1 | 5 | 906 | ||
NM_013882.2 | NP_038910.1 | 4 | 876 | |||
GenBank mRNA | AJ222580.1 | - | 4 | 846 | ||
AK081582.1 | - | 5 | 869 |