Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNQ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204824.1 | NP_001191753.1 | 12 | 1316 | ||
NM_004519.3 | NP_004510.1 | 12 | 1865 | |||
XM_005250914.3 | XP_005250971.1 | 9 | 940 | |||
XM_006716555.3 | XP_006716618.1 | 10 | 1126 | |||
XM_011517026.2 | XP_011515328.1 | 12 | 4399 | |||
XM_017013400.1 | XP_016868889.1 | 12 | 11371 | |||
GenBank mRNA | AB208890.1 | - | 11 | 2368 | ||
AF033347.1 | - | 12 | 1499 | |||
AK296293.1 | - | 12 | 1316 | |||
BC128576.1 | - | 13 | 1608 |