Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128599.1 | NP_001122071.1 | 6, 7 | 1147 | ||
NM_013887.2 | NP_038915.1 | 6, 7 | 1147 | |||
GenBank mRNA | AF147789.1 | - | 6, 7 | 1147 | ||
BC139827.1 | - | 6, 7 | 1088 | 40142922 | ||
DQ278263.1 | - | 2, 3 | 310 | |||
EU303117.1 | - | 6, 7 | 963 | |||
EU303118.1 | - | 6, 7 | 963 |
Set Membership: |
G-protein coupled receptor |