Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253685.1 | NP_001240614.1 | 4 | 368 | ||
NM_013888.3 | NP_038916.1 | 4 | 505 | |||
GenBank mRNA | AF132906.2 | - | 4 | 454 | ||
AK002649.1 | - | 2 | 193 | |||
AK032207.1 | - | 3 | 1874 | |||
BC013501.1 | - | 4 | 412 | 4222828 | ||
BC071211.1 | - | 3 | 1770 | |||
BF453706.1 | - | 4 | 368 | |||
DV042983.1 | - | 4 | 520 |