Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013906.2 | NP_038934.2 | 2 | 1106 | ||
GenBank mRNA | AF175282.1 | - | 2 | 1109 | ||
AK043875.1 | - | 213 |