Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145600.1 | NP_663575.1 | 2 | 151 | ||
GenBank mRNA | AK050534.1 | - | 2 | 161 | ||
AK086617.1 | - | 2 | 185 | |||
BC008086.1 | - | 2 | 151 | 3586161 | ||
BC030675.1 | - | 2 | 149 | 1068780 | ||
BC083081.1 | - | 2 | 185 | 6510396 |