Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152800.3 | NP_690013.1 | 2 | 448 | ||
GenBank mRNA | AK019246.1 | - | 2 | 298 | ||
AK162719.1 | - | 2 | 392 | |||
AK170979.1 | - | 1 | 1627 | |||
AK171247.1 | - | 2 | 400 | |||
BC003466.1 | - | 2 | 433 | 2649388 | ||
BC024469.1 | - | 2 | 411 | 4976022 | ||
CK031473.1 | - | 2 | 309 |