Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133673.3 | NP_598434.2 | 5 | 980 | ||
GenBank mRNA | AJ297743.1 | - | 5 | 1010 | ||
AK017290.1 | - | 5 | 999 | |||
AK028859.1 | - | 5 | 795 | |||
AK035602.1 | - | 1 | 162 | |||
AK054404.1 | - | 5 | 1011 | |||
AK149754.1 | - | 5 | 1001 | |||
AK159843.1 | - | 5 | 979 | |||
AK169692.1 | - | 4 | 2764 | |||
AK171756.1 | - | 3 | 2006 | |||
BC018456.1 | - | 5 | 979 | 3582246 |