Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144799.2 | NP_659048.1 | 4 | 885 | ||
GenBank mRNA | AK075847.1 | - | 4 | 882 | ||
AK159946.1 | - | 4 | 836 | |||
BC019124.1 | - | 4 | 836 | 5007525 |