Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015759.2 | NP_056574.2 | 18 | 2236 | ||
GenBank mRNA | AF017369.1 | - | 18 | 2271 | ||
AK157835.1 | - | 2236 | ||||
AK165413.1 | - | 18 | 2646 | |||
BC132340.1 | - | 16 | 2055 | 40130617 | ||
BC137954.1 | - | 16 | 2056 | 8860973 |