Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013919.4 | NP_038947.2 | 11 | 1648 | ||
GenBank mRNA | AF177759.1 | - | 10 | 1447 | ||
AK011148.1 | - | 11 | 1658 | |||
BC021903.1 | - | 11 | 1638 | 3985792 | ||
KF646669.1 | - | 10 | 1166 |