Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KHK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000221.2 | NP_000212.1 | 6 | 1158 | ||
NM_006488.2 | NP_006479.1 | 6 | 1158 | |||
XM_005264294.3 | XP_005264351.1 | 7 | 1301 | |||
XM_005264296.3 | XP_005264353.1 | 6 | 1144 | |||
XM_005264298.3 | XP_005264355.1 | 4 | 911 | |||
XM_017004060.1 | XP_016859549.1 | 7 | 1298 | |||
XM_017004061.1 | XP_016859550.1 | 5 | 1028 | |||
GenBank mRNA | AB209518.1 | - | 1 | 2779 | ||
AK130033.1 | - | 6 | 675 | |||
AK223075.1 | - | 6 | 675 | |||
BC006233.2 | - | 6 | 1096 | 3946893 | ||
BX648873.1 | - | 4 | 425 | |||
CR456801.1 | - | 6 | 641 | |||
X78677.1 | - | 4 | 437 | |||
X78678.1 | - | 6 | 649 |