Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013925.4 | NP_038953.1 | 9, 10 | 1521 | ||
GenBank mRNA | AF192375.1 | - | 8, 9 | 1360 | ||
AK039699.1 | - | 9, 10 | 1491 | |||
AK082399.1 | - | 9, 10 | 1522 |