Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083334.1 | NP_001076803.1 | 9 | 932 | ||
NM_009668.2 | NP_033798.1 | 10 | 1025 | |||
GenBank mRNA | BC065160.1 | - | 9 | 904 | 5718309 | |
CF732768.1 | - | 1 | 62 | |||
U60884.1 | - | 9 | 920 |