Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025304.3 | NP_079580.2 | 9, 10 | 1037 | ||
GenBank mRNA | AK003424.1 | - | 9, 10 | 962 | ||
AK007326.1 | - | 3, 4 | 286 | |||
AK185158.1 | - | 2, 3 | 141 | |||
BB686087.1 | - | 1, 2 | 81 | |||
BC010316.1 | - | 6, 7 | 490 | |||
BC058379.1 | - | 2, 3 | 1449 | |||
BC132507.1 | - | 9, 10 | 943 | 40130784 | ||
BC132509.1 | - | 9, 10 | 943 | 40130786 | ||
BC145301.1 | - | 9, 10 | 943 |