Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KHK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000221.2 | NP_000212.1 | 8 | 2060 | ||
NM_006488.2 | NP_006479.1 | 8 | 2060 | |||
XM_005264294.3 | XP_005264351.1 | 9 | 2203 | |||
XM_005264296.3 | XP_005264353.1 | 8 | 2046 | |||
XM_005264298.3 | XP_005264355.1 | 6 | 1813 | |||
XM_017004060.1 | XP_016859549.1 | 9 | 2200 | |||
XM_017004061.1 | XP_016859550.1 | 7 | 1930 | |||
GenBank mRNA | AK130033.1 | - | 8 | 1577 | ||
BX648873.1 | - | 6 | 1327 | |||
X78678.1 | - | 8 | 1551 |