Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146200.1 | NP_001139672.1 | 7 | 2747 | ||
NM_001146201.1 | NP_001139673.1 | 7 | 2803 | |||
NM_020272.2 | NP_064668.2 | 7 | 2924 | |||
GenBank mRNA | AJ249280.1 | - | 7 | 2924 | ||
AK040543.1 | - | 7 | 2709 | |||
AK077772.1 | - | 1 | 79 | |||
AK132553.1 | - | 7 | 2749 | |||
AK137470.1 | - | 7 | 2656 | |||
AK137620.1 | - | 7 | 2805 | |||
AK143238.1 | - | 4 | 428 | |||
AY831679.1 | - | 3 | 288 | |||
BC051246.1 | - | 7 | 2664 | 6514979 |