Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk8ip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163447.1 | NP_001156919.1 | 20 | 2512 | ||
NM_001163448.1 | NP_001156920.1 | 19 | 2470 | |||
NM_001163449.1 | NP_001156921.1 | 21 | 2446 | |||
NM_001163450.1 | NP_001156922.1 | 20 | 2422 | |||
NM_001163451.1 | NP_001156923.1 | 20 | 2419 | |||
NM_001163453.1 | NP_001156925.1 | 19 | 2404 | |||
NM_013931.4 | NP_038959.2 | 20 | 2515 | |||
GenBank mRNA | AB005662.1 | - | 20 | 2402 | ||
AB043123.1 | - | 21 | 2323 | |||
AB043124.1 | - | 20 | 2392 | |||
AB043125.1 | - | 20 | 2389 | |||
AB093282.2 | - | 20 | 2374 | |||
AF178636.1 | - | 19 | 2387 | |||
AF178637.1 | - | 20 | 2508 | |||
AF262046.1 | - | 20 | 2498 | |||
AK039130.1 | - | 2 | 142 | |||
BC060603.1 | - | 21 | 2381 | 5706805 | ||
BC065105.1 | - | 19 | 2394 | 30356817 | ||
BC068312.1 | - | 20 | 2493 | 30100914 | ||
CO433928.1 | - | 2 | 252 |