Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033270.2 | NP_150373.2 | 4 | 747 | ||
GenBank mRNA | AB221552.1 | - | 4 | 508 | ||
AF032131.1 | - | 4 | 725 | |||
AK035116.1 | - | 4 | 744 | |||
AK146792.1 | - | 5 | 809 | |||
AK149061.1 | - | 903 | ||||
AK183289.1 | - | 1 | 97 | |||
AK187122.1 | - | 1 | 92 | |||
BC019166.1 | - | 5 | 809 | 4236246 | ||
BC037656.1 | - | 5 | 690 | 5358554 | ||
BC057929.1 | - | 4 | 735 | 5289475 |
Gene Symbol: Polr1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK180462.1 | - | 172 |