Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015765.2 | NP_056580.2 | 11, 12 | 1323 | ||
GenBank mRNA | AK031489.1 | - | 1262 | |||
AK145937.1 | - | 11, 12 | 1277 | |||
AK153397.1 | - | 11, 12 | 1285 | |||
BC002056.1 | - | 11, 12 | 1251 | 3493119 | ||
BI734146.1 | - | 3, 4 | 426 | |||
U08215.1 | - | 11, 12 | 1289 |