Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015765.2 | NP_056580.2 | 9 | 994 | ||
GenBank mRNA | AK031489.1 | - | 933 | |||
AK145937.1 | - | 9 | 948 | |||
AK153397.1 | - | 9 | 956 | |||
BC002056.1 | - | 9 | 922 | 3493119 | ||
BI734146.1 | - | 1 | 97 | |||
BU505607.1 | - | 6 | 645 | |||
U08215.1 | - | 9 | 954 |