Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037539.2 | NP_001032628.1 | 2 | 229 | ||
NM_015768.2 | NP_056583.1 | 2 | 229 | |||
GenBank mRNA | AF182064.1 | - | 2 | 229 | ||
AF182065.1 | - | 2 | 229 | |||
AF182066.1 | - | 2 | 229 | |||
AF487280.1 | - | 2 | 220 | |||
AK015462.1 | - | 2 | 191 | |||
BC116911.1 | - | 2 | 198 | 40126230 | ||
BC144863.1 | - | 2 | 210 | 9053411 | ||
BY232101.1 | - | 2 | 245 |