Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015769.2 | NP_056584.2 | 2 | 542 | ||
GenBank mRNA | AF189285.1 | - | 2 | 542 | ||
AK018375.1 | - | 2 | 362 | |||
AK043366.1 | - | 2 | 344 | |||
AK138588.1 | - | 2 | 336 | |||
AK149326.1 | - | 2 | 375 | |||
BC026792.1 | - | 2 | 333 | 4911382 | ||
BC051036.1 | - | 1 | 73 |