Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015775.2 | NP_056590.2 | 7 | 855 | ||
GenBank mRNA | AF113596.1 | - | 7 | 725 | ||
AF199362.1 | - | 7 | 736 | |||
AF243500.1 | - | 7 | 725 | |||
AK141350.1 | - | 7 | 830 | |||
AK146047.1 | - | 7 | 739 | |||
AK161173.1 | - | 7 | 849 | |||
BC038393.1 | - | 7 | 694 | 2648878 | ||
BC054348.1 | - | 7 | 855 | 2648889 |