Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025305.2 | NP_079581.1 | 5 | 569 | ||
GenBank mRNA | AK009251.1 | - | 5 | 534 | ||
AK012225.1 | - | 5 | 569 | |||
AK131937.1 | - | 5 | 580 | |||
AK162024.1 | - | 5 | 548 | |||
BC049636.1 | - | 5 | 569 | 6532964 |