Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Postn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198765.1 | NP_001185694.1 | 10, 11 | 1438 | ||
NM_001198766.1 | NP_001185695.1 | 10, 11 | 1438 | |||
NM_015784.3 | NP_056599.1 | 10, 11 | 1438 | |||
GenBank mRNA | AK030756.1 | - | 10, 11 | 1437 | ||
AK154049.1 | - | 10, 11 | 1436 | |||
AY651928.1 | - | 10, 11 | 1408 | |||
BC007141.1 | - | 10, 11 | 1432 | |||
BC031449.1 | - | 10, 11 | 1407 | 4457222 | ||
D13664.1 | - | 10, 11 | 1420 | |||
E05655.1 | - | 10, 11 | 1434 | |||
KM117171.1 | - | 10, 11 | 1402 | |||
KM117172.1 | - | 10, 11 | 1402 |