Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015787.4 | NP_056602.1 | 1 | 670 | ||
GenBank mRNA | BC089600.1 | - | 670 | 6745595 |